2-(2-Bromoacetyl)thiophene ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-1-(thiophen-2-yl)ethanone2-(Bromoacetyl)thiophene

081088-1G
102834-012EA 73.85 USD
102834-012
2-(2-Bromoacetyl)thiophene ≥95%
2-(2-Bromoacetyl)thiophene

Hazard: Corrosive/Moisture Sensitive/Refrigerate/Lachrymator

Formula: C₆H₅BrOS
MW: 205.07 g/mol
Melting Pt: 29…33 °C
MDL Number: MFCD02677721
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR