ω-Bromo-2,5-dichloroacetophenone ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-1-(2,5-dichlorophenyl)ethanone

081084-1G
102833-998EA 180.55 USD
102833-998
ω-Bromo-2,5-dichloroacetophenone ≥97%
ω-Bromo-2,5-dichloroacetophenone

Hazard: Irritant

Formula: C₈H₅BrCl₂O
CAS Number: 4571-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR