4'-tert-Butyl-2-chloroacetophenone ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(tert-Butyl)phenacyl chloride

081083-1G
102833-996EA 117.3 USD
102833-996
4'-tert-Butyl-2-chloroacetophenone ≥97%
4'-tert-Butyl-2-chloroacetophenone

Hazard: Irritant

Formula: C₉H₉ClO
MW: 210.71
MDL Number: MFCD00171621
CAS Number: 21886-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR