2-Bromo-3',5'-di-tert-butyl-4'-hydroxyacetophenone ≥97%
Supplier: Matrix Scientific
Synonyms:
3,5-Bis(tert-butyl)-4-hydroxyphenacyl bromide
, 2-Bromo-1-[3,5-di(tert-butyl)-4-hydroxyphenyl]ethan-1-one
081079-1G
081079-10G
102833-986EA
707.69
USD
102833-986
102833-984
2-Bromo-3',5'-di-tert-butyl-4'-hydroxyacetophenone ≥97%
2-Bromo-3',5'-di-tert-butyl-4'-hydroxyacetophenone
Hazard: Irritant
Formula:
C₁₆H₂₃BrO₂ MW: 327.26 g/mol |
MDL Number:
MFCD00173889 CAS Number: 14386-64-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...