Cyclopentylacetyl chloride

Supplier: Matrix Scientific

081019-5G 081019-1G 081019-500MG
102672-418EA 623.08 USD
102672-418 102672-416 102672-414
Cyclopentylacetyl chloride
Cyclopentylacetyl chloride

Hazard: Irritant

Formula: C₇H₁₁ClO
Boiling Pt: 60 °C (12 Torr)
MDL Number: MFCD00236031
CAS Number: 1122-99-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR