N-(Cyanomethyl)acetamide
Supplier: Matrix Scientific
Synonyms:
N-(cyanomethyl)acetamide
081008-500MG
081008-1G
102672-354EA
646.15
USD
102672-354
102672-352
N-(Cyanomethyl)acetamide
N-(Cyanomethyl)acetamide
Hazard: Irritant
Formula:
C₄H₆N₂O MW: 98.11 g/mol |
MDL Number:
MFCD00019815 CAS Number: 4814-80-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...