2-(3,5-Dimethyl-1H-pyrazol-1-yl)quinolin-8-amine
Supplier: Matrix Scientific
080671-1G
080671-5G
080671-500MG
102670-696EA
243.08
USD
102670-696
102670-694
102670-698
2-(3,5-Dimethyl-1H-pyrazol-1-yl)quinolin-8-amine
2-(3,5-Dimethyl-1H-pyrazol-1-yl)quinolin-8-amine
Hazard: Irritant
Formula:
C₁₄H₁₄N₄ |
MDL Number:
MFCD01246359 CAS Number: 356522-39-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...