Ethyl-2-amylacetoacetate
Supplier: Matrix Scientific
Synonyms:
2-Acetylheptanoic Acid Ethyl Ester
, 2-Amylacetoacetic Acid Ethyl Ester
, Ethyl 2-Acetylheptanoate
080547-5G
080547-1G
080547-500MG
102670-094EA
243.08
USD
102670-094
102670-092
102670-096
Ethyl-2-amylacetoacetate
Ethyl-2-amylacetoacetate
CAS: 24317-94-0
MDL Number: MFCD00059380
Molecular Formula: C11H20O3
Molecular Weight: 200,28
Hazard: Irritant
Formula:
C₁₁H₂₀O₃ MW: 200.28 g/mol |
MDL Number:
MFCD00059380 CAS Number: 24317-94-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...