Methyl-3-cyclopropyl-1H-pyrazole-5-carboxylate
Supplier: Matrix Scientific
Synonyms:
Methyl 5-cyclopropyl-1H-pyrazole-3-carboxylate
080276-500MG
080276-5G
102668-816EA
1615.38
USD
102668-816
102668-812
Methyl-3-cyclopropyl-1H-pyrazole-5-carboxylate
Methyl-3-cyclopropyl-1H-pyrazole-5-carboxylate
Hazard: Irritant
Formula:
C₈H₁₀N₂O₂ MW: 166.18 g/mol |
MDL Number:
MFCD08559008 CAS Number: 1036733-11-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...