2-Quinolinecarbaldehyde

Supplier: Matrix Scientific

Synonyms: Quinoline-2-carboxaldehydeQuinoline-2-carbaldehyde2-Quinolinecarboxaldehyde

080103-25G
102833-964EA 258.46 USD
102833-964
2-Quinolinecarbaldehyde
2-Quinolinecarbaldehyde

Hazard: Irritant/Air Sensitive/Stench

Formula: C₁₀H₇NO
MW: 157.17 g/mol
Melting Pt: 70…72 °C
MDL Number: MFCD00075032
CAS Number: 5470-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR