1-(2-Aminophenyl)pyrrole

Supplier: Matrix Scientific

Synonyms: 2-(1H-Pyrrol-1-yl)aniline

079047-1G
102668-392EA 484.62 USD
102668-392
1-(2-Aminophenyl)pyrrole
1-(2-Aminophenyl)pyrrole

Hazard: Irritant

Formula: C₁₀H₁₀N₂
MW: 158.20 g/mol
MDL Number: MFCD00005344
CAS Number: 6025-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR