2-Bromo-N-methylbenzamide

Supplier: Matrix Scientific

077871-1G
102668-128EA 144.9 USD
102668-128
2-Bromo-N-methylbenzamide
2-Bromo-N-methylbenzamide

Hazard: Irritant

Formula: C₈H₈BrNO
MW: 214.06 g/mol
MDL Number: MFCD00099250
CAS Number: 61436-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR