5-Phenylazosalicylaldehyde

Supplier: Matrix Scientific

077851-1G
102668-038EA 484.62 USD
102668-038
5-Phenylazosalicylaldehyde
5-Phenylazosalicylaldehyde

Hazard: Irritant

Formula: C₁₃H₁₀N₂O₂
MDL Number: MFCD00445643
CAS Number: 151726-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR