2-amino-3-chloropropanoic acid hydrochloride

Supplier: Matrix Scientific

Synonyms: H-β-Chloro-DL-Ala-OH·HCl

077833-1G
102667-974EA 646.15 USD
102667-974
2-amino-3-chloropropanoic acid hydrochloride
2-amino-3-chloropropanoic acid hydrochloride

Hazard: Irritant

Formula: C₃H₇Cl₂NO₂
MW: 159.994 g/mol
MDL Number: MFCD00066125
CAS Number: 35401-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR