3-Amino-5-methylthio-1H-1,2,4-triazole

Supplier: Matrix Scientific

Synonyms: 3-Amino-5-(methylmercapto)-1H-1,2,4-triazole

077770-1G
102667-660EA 144.9 USD
102667-660
3-Amino-5-methylthio-1H-1,2,4-triazole
3-Amino-5-methylthio-1H-1,2,4-triazole

Hazard: Irritant

Formula: C₃H₆N₄S
MDL Number: MFCD00005232
CAS Number: 45534-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR