6,7-Dihydroimidazo[1,2-a]pyridin-8(5H)-one
Supplier: Matrix Scientific
077615-1G
077615-5G
077615-500MG
102667-122EA
775.38
USD
102667-122
102667-124
102667-126
6,7-Dihydroimidazo[1,2-a]pyridin-8(5H)-one
6,7-Dihydroimidazo[1,2-a]pyridin-8(5H)-one
Hazard: Irritant
Formula:
C₇H₈N₂O |
MDL Number:
MFCD18830033 CAS Number: 457949-09-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...