N-(2-Hydroxypropyl)morpholine
Supplier: Matrix Scientific
Synonyms:
1-Morpholino-2-propanol
077536-500MG
077536-1G
077536-5G
102666-764EA
516.92
USD
102666-764
102666-768
102666-766
N-(2-Hydroxypropyl)morpholine
N-(2-Hydroxypropyl)morpholine
CAS: 2109-66-2
MDL Number: MFCD00023378
Molecular Formula: C7H15NO2
Molecular Weight: 145,2
Hazard: Irritant
Formula:
C₇H₁₅NO₂ MW: 145.20 g/mol |
MDL Number:
MFCD00023378 CAS Number: 2109-66-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...