2-Acetyl-4-methylpyridine ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(4-Methylpyridin-2-yl)ethanone

077073-1G
102664-374EA 38.46 USD
102664-374
2-Acetyl-4-methylpyridine ≥97%
2-Acetyl-4-methylpyridine

Hazard: Irritant

Formula: C₈H₉NO
MW: 135.1632 g/mol
Melting Pt: 28…33 °C
MDL Number: MFCD01863638
CAS Number: 59576-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR