5,6-Dichloropicolinic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 5,6-Dichloropyridine-2-carboxylic acid

077056-1G
102664-348EA 136.92 USD
102664-348
5,6-Dichloropicolinic acid ≥98%
5,6-Dichloropicolinic acid

Hazard: Irritant

Formula: C₆H₃Cl₂NO₂
MW: 192.00 g/mol
MDL Number: MFCD13185546
CAS Number: 88912-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR