4-Formyl-2-picoline ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Methylisonicotinaldehyde

076651-500MG 076651-1G
102547-280EA 135.38 USD
102547-280 102547-282
4-Formyl-2-picoline ≥97%
4-Formyl-2-picoline

Hazard: Irritant

Formula: C₇H₇NO
MW: 121.1400 g/mol
MDL Number: MFCD06410683
CAS Number: 63875-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR