3-Bromo-5-(trifluoromethyl)phenol ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Bromo-5-hydroxybenzotrifluoride
076607-1G
076607-5G
102547-080EA
60
USD
102547-080
102547-078
3-Bromo-5-(trifluoromethyl)phenol ≥95%
3-Bromo-5-(trifluoromethyl)phenol
Hazard: Irritant
Formula:
C₇H₄BrF₃O MW: 241.0100 g/mol |
MDL Number:
MFCD11041083 CAS Number: 1025718-84-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...