2-Ethylthiophene ≥95%

Supplier: Matrix Scientific

Synonyms: o-Ethylthiophene

076586-5G
102546-972EA 72.31 USD
102546-972
2-Ethylthiophene ≥95%
2-Ethylthiophene

Hazard: Irritant

Formula: C₆H₈S
MW: 112.2 g/mol
MDL Number: MFCD00005461
CAS Number: 872-55-9
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR