2-Bromo-5-(trifluoromethyl)anisole ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-3-methoxybenzotrifluoride

076558-1G 076558-5G
102546-826EA 39.1 USD
102546-826 102546-828
2-Bromo-5-(trifluoromethyl)anisole ≥95%
2-Bromo-5-(trifluoromethyl)anisole

Hazard: Irritant/Refrigerate

Formula: C₈H₆BrF₃O
MW: 255.03 g/mol
Melting Pt: 26…28 °C
MDL Number: MFCD04115974
CAS Number: 402-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR