1-(3-Nitrophenyl)piperazine ≥95%
Supplier: Matrix Scientific
076542-5G
076542-10G
076542-1G
102546-748EA
463.08
USD
102546-748
102546-746
102546-750
1-(3-Nitrophenyl)piperazine ≥95%
1-(3-Nitrophenyl)piperazine
Hazard: Irritant
Formula:
C₁₀H₁₃N₃O₂ MW: 207.23 g/mol |
CAS Number:
54054-85-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...