5-Bromopicolinamide ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Bromopyridine-2-carboxamide
076499-5G
076499-1G
102546-524EA
426.15
USD
102546-524
102546-522
5-Bromopicolinamide ≥95%
5-Bromopicolinamide
Hazard: Irritant
Formula:
C₆H₅BrN₂O MW: 201.0200 g/mol |
MDL Number:
MFCD04066715 CAS Number: 90145-48-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...