3-Formyl-2-picoline ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Methylnicotinaldehyde

076491-5G 076491-10G 076491-1G
102546-480EA 1147.7 USD
102546-480 102546-482 102546-478
3-Formyl-2-picoline ≥95%
3-Formyl-2-picoline

Hazard: Irritant

Formula: C₇H₇NO
MDL Number: MFCD08234911
CAS Number: 60032-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR