Ethyl-5-methyl-1H-imidazole-4-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 5-methyl-1H-imidazole-4-carboxylate
, 4-Methyl-1H-imidazole-5-carboxylic acid ethyl ester
, Ethyl 4-methyl-5-imidazolecarboxylate
, Ethyl 4-Methyl-1H-imidazole-5-carboxylate
076488-25G
076488-5G
076488-1G
102833-924EA
973.85
USD
102833-924
102546-462
102546-460
Ethyl-5-methyl-1H-imidazole-4-carboxylate ≥95%
Ethyl-5-methyl-1H-imidazole-4-carboxylate
Hazard: Irritant
Formula:
C₇H₁₀N₂O₂ MW: 154.1665 g/mol |
MDL Number:
MFCD00005199 CAS Number: 51605-32-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...