(R)-(+)-4-Bromo-α-methylbenzylamine ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-1-(4-Bromophenyl)ethylamine(R)-(+)-1-(4-Bromophenyl)ethylamine

076487-5G
102546-456EA 93.85 USD
102546-456
(R)-(+)-4-Bromo-α-methylbenzylamine ≥95%
(R)-(+)-4-Bromo-α-methylbenzylamine

Hazard: Corrosive/Air Sensitive

Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Boiling Pt: 140…145 °C (30 mmHg)
Density: 1.371
MDL Number: MFCD00025548
CAS Number: 45791-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR