2-Methylmorpholine ≥95%
Supplier: Matrix Scientific
076484-1G
102546-436EA
241.54
USD
102546-436
2-Methylmorpholine ≥95%
2-Methylmorpholine
Hazard: Irritant
Formula:
C₅H₁₁NO MW: 101.1469 g/mol |
MDL Number:
MFCD08444368 CAS Number: 27550-90-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...