2-Methylthiophene-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

076477-1G 076477-5G 076477-25G
102546-396EA 44.62 USD
102546-396 102546-398 102552-032
2-Methylthiophene-3-carboxylic acid ≥95%
2-Methylthiophene-3-carboxylic acid

Hazard: Irritant

Formula: C₆H₆O₂S
MW: 142.1756 g/mol
MDL Number: MFCD01859891
CAS Number: 1918-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR