6-Hydroxy-7-methoxyquinazolin-4(1H)-one ≥95%
Supplier: Matrix Scientific
Synonyms:
6-Hydroxy-7-methoxy-4(3H)-quinazolinone
, 6-Hydroxy-7-methoxy-3H-quinazolin-4-one
076474-5G
102546-382EA
186.15
USD
102546-382
6-Hydroxy-7-methoxyquinazolin-4(1H)-one ≥95%
6-Hydroxy-7-methoxyquinazolin-4(1H)-one
Hazard: Irritant
Formula:
C₉H₈N₂O₃ MW: 192.1714 g/mol |
MDL Number:
MFCD20547768 CAS Number: 179688-52-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...