3-Fluoro-2-iodoaniline ≥95%

Supplier: Matrix Scientific

076472-5G 076472-250MG 076472-1G
102546-370EA 1383.08 USD
102546-370 102546-372 102546-368
3-Fluoro-2-iodoaniline ≥95%
3-Fluoro-2-iodoaniline

Hazard: Irritant

Formula: C₆H₅FIN
MW: 237.0100 g/mol
MDL Number: MFCD08458368
CAS Number: 706752-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR