3-Chloro-2-methylpyridine ≥95%
Supplier: Matrix Scientific
076470-1G
076470-5G
102546-356EA
369.23
USD
102546-356
102546-358
3-Chloro-2-methylpyridine ≥95%
3-Chloro-2-methylpyridine
Hazard: Irritant/Irritant-Harmful
Formula:
C₆H₆ClN MW: 127.5700 g/mol Boiling Pt: 156.5 °C |
MDL Number:
MFCD04114121 CAS Number: 72093-03-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...