Methyl-2-amino-1H-benzo[d]imidazole-5-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 2-amino-1H-benzo[d]imidazole-5-carboxylate
076468-250MG
076468-1G
102546-348EA
2087.69
USD
102546-348
102546-346
Methyl-2-amino-1H-benzo[d]imidazole-5-carboxylate ≥95%
Methyl-2-amino-1H-benzo[d]imidazole-5-carboxylate
Hazard: Irritant
Formula:
C₉H₉N₃O₂ MW: 191.1900 g/mol |
MDL Number:
MFCD08752985 CAS Number: 106429-38-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...