2-Hydroxythiazole ≥95%

Supplier: Matrix Scientific

076426-1G 076426-250MG 076426-5G
102546-120EA 413.85 USD
102546-120 102546-122 102546-118
2-Hydroxythiazole ≥95%
2-Hydroxythiazole

Hazard: Irritant

Formula: C₃H₃NOS
MW: 101.1270 g/mol
MDL Number: MFCD02094166
CAS Number: 82358-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR