tert-Butyl (2-bromothiazol-4-yl)carbamate ≥95%
Supplier: Matrix Scientific
076380-5G
076380-250MG
076380-1G
102545-876EA
815.38
USD
102545-876
102545-872
102545-874
tert-Butyl (2-bromothiazol-4-yl)carbamate ≥95%
tert-Butyl (2-bromothiazol-4-yl)carbamate
Hazard: Irritant
Formula:
C₈H₁₁BrN₂O₂S MW: 279.1541 g/mol |
MDL Number:
MFCD12032212 CAS Number: 1245647-95-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...