3-Amino-2,6-dimethylpyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
2,6-Dimethylpyridin-3-amine
076377-5G
076377-1G
076377-10G
102545-858EA
56.92
USD
102545-858
102545-856
102545-860
3-Amino-2,6-dimethylpyridine ≥95%
3-Amino-2,6-dimethylpyridine
Hazard: Irritant
Formula:
C₇H₁₀N₂ MW: 122.17 g/mol |
MDL Number:
MFCD01646409 CAS Number: 3430-33-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...