4-Bromo-5-fluoro-2-methylbenzaldehyde ≥95%

Supplier: Matrix Scientific

076374-1G 076374-5G
102545-842EA 853.85 USD
102545-842 102545-840
4-Bromo-5-fluoro-2-methylbenzaldehyde ≥95%
4-Bromo-5-fluoro-2-methylbenzaldehyde

Hazard: Irritant

Formula: C₈H₆BrFO
MW: 217.0400 g/mol
MDL Number: MFCD07782049
CAS Number: 861928-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR