5-Bromo-4-methoxythiophene-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

076369-10G 076369-1G 076369-5G
102545-814EA 1996.92 USD
102545-814 102545-816 102545-812
5-Bromo-4-methoxythiophene-3-carboxylic acid ≥95%
5-Bromo-4-methoxythiophene-3-carboxylic acid

Hazard: Irritant

Formula: C₆H₅BrO₃S
CAS Number: 162848-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR