(S)-(-)-3-Chloro-1-phenyl-1-propanol ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-3-Chloro-1-phenylpropan-1-ol

076365-5G
102545-790EA 207.69 USD
102545-790
(S)-(-)-3-Chloro-1-phenyl-1-propanol ≥95%
(S)-(-)-3-Chloro-1-phenyl-1-propanol

Hazard: Irritant

Formula: C₉H₁₁ClO
MW: 170.64 g/mol
MDL Number: MFCD00013309
CAS Number: 100306-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR