1-Chloroisoquinoline ≥95%
Supplier: Matrix Scientific
076363-10G
076363-5G
076363-1G
102545-780EA
183.08
USD
102545-780
102545-776
102545-778
1-Chloroisoquinoline ≥95%
1-Chloroisoquinoline
Hazard: Irritant
Formula:
C₉H₆ClN MW: 163.61 g/mol |
MDL Number:
MFCD00024134 CAS Number: 19493-44-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...