2-Methoxy-5-nitrophenol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Hydroxy-4-nitroanisole5-Nitroguaiacol

076348-100G
102545-700EA 110.77 USD
102545-700
2-Methoxy-5-nitrophenol ≥95%
2-Methoxy-5-nitrophenol

Hazard: Irritant

Formula: C₇H₇NO₄
MW: 169.14 g/mol
MDL Number: MFCD00015561
CAS Number: 636-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR