6-[3-(Trifluoromethyl)phenoxy]-2-pyridinecarboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
6-(3-(Trifluoromethyl)phenoxy)picolinic acid
076320-250MG
076320-1G
102545-568EA
2640
USD
102545-568
102545-566
6-[3-(Trifluoromethyl)phenoxy]-2-pyridinecarboxylic acid ≥95%
6-[3-(Trifluoromethyl)phenoxy]-2-pyridinecarboxylic acid
Hazard: Irritant
Formula:
C₁₃H₈F₃NO₃ MW: 283.2000 g/mol |
MDL Number:
MFCD00227526 CAS Number: 137640-84-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...