1-Methylcyclopropane-1-sulfonamide ≥95%
Supplier: Matrix Scientific
076294-1G
076294-5G
102545-436EA
370.77
USD
102545-436
102545-434
1-Methylcyclopropane-1-sulfonamide ≥95%
1-Methylcyclopropane-1-sulfonamide
Hazard: Irritant
Formula:
C₄H₉NO₂S MW: 135.1900 g/mol |
MDL Number:
MFCD11501466 CAS Number: 669008-26-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...