5-Ethyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Ethyl-2-thiouracil

076293-1G 076293-5G
102545-430EA 533.85 USD
102545-430 102545-432
5-Ethyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one ≥95%
5-Ethyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one

Hazard: Irritant

Formula: C₆H₈N₂OS
MW: 156.2100 g/mol
MDL Number: MFCD00079592
CAS Number: 34171-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR