3-Methoxy-5-nitrobenzoic acid ≥95%
Supplier: Matrix Scientific
076277-1G
076277-5G
102545-350EA
1841.54
USD
102545-350
102545-348
3-Methoxy-5-nitrobenzoic acid ≥95%
3-Methoxy-5-nitrobenzoic acid
Hazard: Irritant
Formula:
C₈H₇NO₅ MW: 197.1500 g/mol |
MDL Number:
MFCD01318045 CAS Number: 78238-12-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...