1-Amino-2-(isopropylsulfonyl)benzene ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Isopropylsulphonyl)aniline2-(Isopropylsulfonyl)aniline2-Aminophenyl Isopropyl sulphone2-Aminophenyl Isopropyl sulfone

076276-1G
102545-346EA 198.46 USD
102545-346
1-Amino-2-(isopropylsulfonyl)benzene ≥95%
1-Amino-2-(isopropylsulfonyl)benzene

Hazard: Irritant

Formula: C₉H₁₃NO₂S
MW: 199.2700 g/mol
MDL Number: MFCD08445603
CAS Number: 76697-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR