3-Methyl-7-propylxanthine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Methyl-7-propyl-3,7-dihydro-1H-purine-2,6-dione

076272-1G 076272-250MG 076272-5G
102545-326EA 921.54 USD
102545-326 102545-328 102545-324
3-Methyl-7-propylxanthine ≥95%
3-Methyl-7-propylxanthine

Hazard: Irritant

Formula: C₉H₁₂N₄O₂
MW: 208.2172 g/mol
MDL Number: MFCD02089337
CAS Number: 55242-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR