5-Chloro-2-methoxy-4-pyridineboronic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Chloro-2-methoxypyridine-4-boronic acid
, (5-Chloro-2-methoxypyridin-4-yl)boronic acid
076269-5G
076269-1G
102545-310EA
98.46
USD
102545-310
102545-312
5-Chloro-2-methoxy-4-pyridineboronic acid ≥95%
5-Chloro-2-methoxy-4-pyridineboronic acid
Hazard: Irritant/Refrigerate
Formula:
C₆H₇BClNO₃ MW: 187.3900 g/mol |
MDL Number:
MFCD06658398 CAS Number: 475275-69-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...