2-Amino-4-(trifluoromethyl)benzonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Amino-4-cyanobenzotrifluoride

076263-5G
102545-274EA 178.46 USD
102545-274
2-Amino-4-(trifluoromethyl)benzonitrile ≥95%
2-Amino-4-(trifluoromethyl)benzonitrile

Hazard: Irritant

Formula: C₈H₅F₃N₂
MW: 186.1339 g/mol
MDL Number: MFCD04974124
CAS Number: 1483-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR